Would you like to log in again?
KIMBLE® Large-Volume Vacuum Traps with O-Ring Joint, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE®
926055-0500
KT926055-0500EA 566.28 USD InStock
KT926055-0500
KIMBLE® Large-Volume Vacuum Traps with O-Ring Joint, DWK Life Sciences
Vacuum Traps
This two-piece trap is mainly designed for large volume solvent removal by vacuum.
  • Manufactured from 33 expansion, low extractable borosilicate glass
  • Connections accept standard flexible ¹/₂" hose
The presence of O-ring joint provides a vacuum-tight, grease-free seal which permits the trap body to be easily disconnected from the stopper for cleaning.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty