Would you like to log in again?
KIMBLE® ULTRA-WARE® Four Valve Filtration Delivery Cap, THF Resitant, DWK Life Sciences
Supplier: DWK Life Sciences
ULTRA-WARE®
953981-6347
KT953981-6347EA 2683.18 USD InStock
KT953981-6347
KIMBLE® ULTRA-WARE® Four Valve Filtration Delivery Cap, THF Resitant, DWK Life Sciences
Closures Bottle Closures
Tetrahydrofuran (THF), widely used as a mobile phase for GPC, attacks the CTFE valve stems and TFE/propylene O-rings used in the standard four valve cap.
  • This special THF-resistant version has 316 stainless steel wherever the standard system has CTFE and FFKM O-rings in place of the TFE/propylene O-rings
  • This cap is also recommended for aggressive organic solutions such as chlorinated hydrocarbons, ethers and ketones
  • Filter diameter of 47 mm
Cap body is manufactured from glass-filled PTFE, and will fit any reservoir or bottle with a GL 45 thread.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty