Would you like to log in again?
EMS Premium Economy Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78326-76 78326-85 78326-73 78326-74 78326-83 78326-75 78326-80 78326-79 78326-86 78326-88 78326-77 78326-70 78326-72 78326-89 78326-71 78326-81 78326-87 78326-78 78326-82 78326-84
76376-344EA 25.27 USD InStock
76376-344 76376-322 76376-332 76376-310 76376-324 76376-312 76376-334 76376-326 76376-336 76376-314 76376-306 76376-328 76376-316 76376-338 76376-340 76376-342 76376-320 76376-330 76376-308 76376-318
EMS Premium Economy Tweezers, Electron Microscopy Sciences
Forceps
Premium economy tweezers made of 316 stainless steel.
  • Inexpensive tweezers beneficial in non-critical applications
  • Tweezers are anti-acid/non-corrosive and anti-magnetic
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty