Would you like to log in again?
KIMBLE® Propeller Combination Shaft, DWK Life Sciences
Supplier: DWK Life Sciences
790015-0000
KT790015-0000EA 110.77 USD InStock
KT790015-0000
KIMBLE® Propeller Combination Shaft, DWK Life Sciences
Stirrers Stirring Shafts
Propeller combination shaft made from #303 stainless steel.
  • Features three angled blades
  • Shaft diameter is ³/₈"
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty