Would you like to log in again?
PFA FAST SC Autosampler Probe, 1.0 mm int.Ø., PerkinElmer
Supplier: PerkinElmer
N0777522
10139-488EA 1509.06 USD InStock
10139-488
PFA FAST SC Autosampler Probe, 1.0 mm int.Ø., PerkinElmer
Autosamplers
PFA FAST probe offers high chemical resistance for applications involving strong acids, alkalis and organics.
  • Contoured design reduces carryover
  • PFA material provides a high chemical resistance
  • Carbon fiber support
  • Compatible with SC-8 and SC-14 autosamplers
A contoured design reduces carryover, while an all PFA wetted sample flow path reduces contamination. SC Autosampler Probe has a carbon fiber support, 1.0 mm I.D. (gray marker), 150 cm length, connects to port #5 (1/4-28 fitting) high flow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty