Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Rectangular Basket, Marlin Steel Wire Products
Supplier: Marlin Steel Wire Products
00-00007001A-31
76338-088EA 678.1 USD InStock
76338-088
Rectangular Basket, Marlin Steel Wire Products
Baskets Wire Baskets
Rectangular material handling basket with handles and mesh
  • Electropolished finish
  • With four mesh openings per linear inch
  • Handles for easy gripping
  • Stacks so that parts inside the basket do not get crushed
Robust quality that is ideal for washing parts, parts degreasing, ultrasonic cleaning, curing, and materials handling in a variety of applications such as chemical bath, heat treating, and cryogenics.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty