Would you like to log in again?
Radioactive I Labels, Labelmaster
Supplier: Labelmaster
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
HML14 HMSL14
101376-364PK 74.4 USD InStock
101376-364 101377-912
Radioactive I Labels, Labelmaster
Labels
Ship with confidence when using Worded Radioactive I labels, revised to meet the design requirements of 49 CFR 172.407.
  • Printed with light-fast inks for fade resistance
  • Adhesives are formulated for use in various climates and environments
  • Once affixed, labels won't fade or fall off
  • Outstanding chemical and abrasion resistance
  • White and black
In addition to complying with directives set by the DOT, these labels also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty