Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium thiosulfate 0.03148 Normal

Supplier: Ricca Chemical
Warning

R7913000-20F
RCR7913000-20FEA 208.41 USD InStock
RCR7913000-20F
Sodium thiosulfate 0.03148 Normal
Sodium thiosulphate pentahydrate


Formula: Na₂S₂O₃.5H₂O
MW: 248.19 g/mol
Boiling Pt: 100°C
Melting Pt: 0.0°C
Density: 1.0050
MDL Number: MFCD00149186
CAS Number: 10102-17-7
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Potassium Iodate/Starch) 0.03146-0.03150 N at 20°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty