Would you like to log in again?
Special Permit Packaging DOT-SP-9168 Bottle Kits, Labelmaster
Supplier: Labelmaster
HMS-500 HMS-08
103258-542EA 28.4 USD InStock
103258-542 103258-540
Special Permit Packaging DOT-SP-9168 Bottle Kits, Labelmaster
Mailers and Shippers
The fully assembled Special Permit Packaging DOT-SP 9168 Bottle Kits can be used to ship Packing Groups I, II, or III materials by all modes of transport.
  • Meets Procedure 1A requirements
  • Accepted by UPS and FedEx as an exemption package
  • Multiple bottle sizes available
The exemption packages can be used to ship solids or liquid materials with a specific gravity of 2.0 or less. The DOT-SP 9168 kits carry an exemption that allows shipment of Hazard Classes 3, 4, 5, 6, 8, and 9 domestically without using a hazard class label.
Delivery information: Kit contains PVC coated glass bottle with Teflon®-lined cap, full friction tinplate steel canister with welded side seam and lid, UN marked fiberboard double-wall box, absorbent pouch, fiber core (for narrow mouth container), and 4 mil polyethylene bag.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty