Would you like to log in again?
BioClean™ Prelude BPSL Sterile Latex Cleanroom Gloves, Ansell
Supplier: Ansell
BioClean™
BPSL-8085 BPSL-5055 BPSL-7075 BPSL-6065
75982-178CS 425.7 USD InStock
75982-178 75982-176 75982-180 75982-174
BioClean™ Prelude BPSL Sterile Latex Cleanroom Gloves, Ansell
Gloves
Designed for easy double-donning the BioClean™ prelude sterile latex cleanroom gloves provide exceptional flexibility and comfort for increased dexterity and prolonged use.
  • Easy double-donning
These natural colored 305 mm (12") long, latex gloves feature a textured surface for enhanced grip and a beaded cuff for stability on the arm.
Certifications: EN 455 Part 2. EN 420:2003 + A1:2009; EN 420:2003 + A1:2009. ISO 9001:2015; ISO 9001:2008. Category III.
Packaging: One pair per inner PE wallet; one wallet per sealed non-particulating EasyTear™ PE pouch; 10 pouches per sealed outer PE bag; 20 outer bags per lined carton (200 pairs).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty