Would you like to log in again?
MICROCHEM® AlphaTec® 68-2000 Chemical Barrier Coveralls, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Microchem® by AlphaTec®
WH20-B-92-107-05
76020-680CS 185.85 USD InStock
76020-680
MICROCHEM® AlphaTec® 68-2000 Chemical Barrier Coveralls, Ansell
Disposable Garments
Superior breathable microporous laminate technology offering Type 5/6 protection.
  • Proven protective barrier to low concentration liquid chemicals
  • Moisture vapor permeable fabric ('breathable') to help reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Anti-static
  • Low linting to help reduce the risk of cross contamination in critcal areas
  • Silicone-free, critcal in spray painting applications
MICROCHEM® by AplhaTec™ 2000 coveralls provides both protection and comfort with exceptional liquid and particulate protection. Ideal for a wide range of industrial applications.
Certifications: ASTM F1671. Tested according to EN 1149-5 and AATCC 76.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty