Would you like to log in again?
AlphaTec® 2000 Overshoes, Ansell
Supplier: Ansell
AlphaTec®
WH20-B-92-407-00
76311-694CS 891.56 USD InStock
76311-694
AlphaTec® 2000 Overshoes, Ansell
Disposable Garments
Superior breathable microporous laminate technology.
  • Proven protective barrier to low concentration liquid chemicals
  • Optimized fit, improving wearer comfort and safety
  • Low linting
Microporous polyethylene laminate non-woven AlphaTec® 2000 overshoes are designed with elasticated opening, PVC anti-slip sole, and bound seams.
Certifications: CE Category l.
Packaging: Packaged 150 pairs per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty