Would you like to log in again?
Economy Safety Vest, V2CL2MOZ Series, ANSI/ISEA Class 2, MCR Safety
Supplier: MCR Safety
V2CL2MOZX4 V2CL2MOZX3 V2CL2MOZX2
76340-164EA 10.73 USD InStock
76340-164 76340-066 76340-116
Economy Safety Vest, V2CL2MOZ Series, ANSI/ISEA Class 2, MCR Safety
Workwear
This ANSI/ISEA Class 2 value safety vest is made from fluorescent orange polyester mesh material.
  • Zippered front closure
  • 2" silver reflective stripes
  • Fluorescent orange mesh construction
  • No pockets
Mesh material offers increased breathability in warm environments.
Certifications: ANSI/ISEA 107-2015 Class 2, Type R compliant.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty