Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pneumatic Adjustable-Height Standing Desk, 56", Luxor

Supplier: Luxor
SPN56F-BK/TK
76203-766EA 893.92 USD InStock
76203-766
Pneumatic Adjustable-Height Standing Desk, 56", Luxor
Furniture Tables and Workbenches
Quickly deliver collaborative and productive comfort to your workday by seamlessly alternating between sitting and standing with the Luxor pneumatic adjustable height standing desk.

  • Lever-activated pneumatic air cylinder easily adjusts anywhere between 27.5" and 44.5"
  • Constructed with two sturdy steel legs, each reinforced with durable inner legs
  • Chamfered edges keep forearms comfortable while typing and working
  • Sturdy, scratch-resistant desktop with a teak finish
  • Minimal assembly required
  • Meets ANSI/BIFMA standards

Its lever-activated pneumatic air cylinder lets you easily alter the desk's height between 27.5" and 44.5", smoothly lifting your monitors, computer and other office accessories. With two sturdy steel legs, reinforced inner legs and a high-quality laminate finish, this stand-up desk is built to last. With no power outlets required for use, it's perfect for any corporate or home office space.

Overall: W56×D26×H27.5 - 44.5";
Desktop: W56×D26"

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty