Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

12 - Tablet Wall or Desk Charging Box, Luxor

Supplier: Luxor
LLTMW12-G
76203-834EA 729.56 USD InStock
76203-834
12 - Tablet Wall or Desk Charging Box, Luxor
Furniture Storage Cabinets
Perfect for small offices, classrooms and libraries, Luxor wall/desk tablet charging cabinets provide a strong-yet-simple design that's reliably secure yet affordably priced.

  • Charges, stores and transports up to 12 tablets
  • Charging station can be mounted to a wall or sit on any flat surface
  • Padded interior with rubber-coated dividers safeguard devices from damage
  • Secure key-lock; includes two keys
  • Ventilated sides allow constant airflow to avoid overheating
  • Includes UL-Listed 12-outlet 120 V/15 A horizontal electrical power strip
  • Arrives assembled; includes drywall mounting hardware
  • Maximum weight capacity of 30 lbs. (evenly distributed)

Do away with cord clutter while safely ensuring devices are always charged and ready for use. A steel shell with lockable door, coupled with a padded interior and rubber-coated dividers protect your devices, and the compact size makes Luxor Tablet Charging Cabinets great for wall-mount or desktop use.

Divider slots: 1W×11.5"D with 13"H of top clearance;
Cord length: 10'

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty