Would you like to log in again?
WHEATON® French Square Bottles, DWK Life Sciences
Supplier: DWK Life Sciences
W217880 W217883 W217882 W217884
10847-502CS 286.04 USD InStock
10847-502 10847-498 10847-506 10847-504
WHEATON® French Square Bottles, DWK Life Sciences
Bottles Glass Bottles
High-quality bottles are made from clear USP Type III soda-lime glass.
  • Ideal for solvent, chemical, or sample storage
  • Square shape maximizes storage space
  • Available with caps attached or bulk packs without caps
These are ideal for field use or general laboratory use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty