Would you like to log in again?
WHEATON® Headspace Vials, Crimp Top, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
W225279
89495-254CS 59.53 USD InStock
89495-254
WHEATON® Headspace Vials, Crimp Top, DWK Life Sciences
Vials
Crimp top vials manufactured from high-quality clear type I borosilicate glass.
  • Accept 20 mm aluminum seals
  • Variety of aluminum seal styles and septa materials available
  • Purchase vials separately or together with seals in convenience packs
Saves time and reduce the risk of contamination.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty