Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

S100B (Mouse) ELISA Kit, BioVision

Supplier: BioVision

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

AB285283-96TEST
76380-384EA 1177.05 USD InStock
76380-384
S100B (Mouse) ELISA Kit, BioVision
Assays ELISAs
A sandwich ELISA kit for the quantitative measurement of S100B in mouse serum, plasma and other biological fluids.

  • Detection range: 78.13 to 5000 pg/ml
  • Precision: coefficient of variation is <10%
  • Quantitative measurement of S100B in mouse serum, plasma and other biological fluids
  • Sensitivity: 46.88 pg/ml
  • Specificity: This kit recognizes Mouse S100B in samples
  • No significant cross-reactivity or interference between mouse S100B and analogues was observed

S100 proteins regulate a variety of cellular processes such as cell growth and motility, cell cycle progression, transcription, and differentiation. Research studies have demonstrated that significant correlation exists between aberrant S100 protein expression and cancer progression. S100 proteins primarily mediate immune responses in various tissue types but are also involved in neuronal development. S100B (mouse) ELISA kit is based on sandwich-ELISA principle. The micro ELISA plate provided in this kit has been pre-coated with an antibody specific to mouse S100B. Standards or samples are added to the micro ELISA plate wells that bind to the specific antibody. Then a biotinylated detection antibody specific for mouse S100B and Avidin-Horseradish Peroxidase (HRP) conjugate are added successively to each micro plate well and incubated. The wells are washed, a TMB substrate solution is added to the wells and blue color develops in proportion to the amount of S100B bound. The enzyme-substrate reaction is terminated by the addition of stop solution and the color turns yellow. The optical density (OD) is measured spectrophotometrically at a wavelength of 450 ±2 nm. The OD value is proportional to the concentration of mouse S100B. The concentration of mouse S100B in the samples can be calculated by comparing the OD of the samples to the standard curve.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty