Would you like to log in again?
Methacrylated Gelatin
Supplier: Advanced BioMatrix
5215-1KIT
103701-174EA 196.15 USD InStock
103701-174
Methacrylated Gelatin
Proteins and Peptides
PhotoGel®-IRG is a methacrylated gelatin kit. These 3D gelatin hydrogels can be prepared at various concentrations and photocrosslinked (365nm) to provide various gel stiffness. Each kit contains 1 gram (2 x 500 mg) of methacrylated gelatin and irgacure photoinitiator.
  • Highly purified
  • Lyophilized to allow you to choose the desired concentration
Advanced BioMatrix offers PhotoGel® UV Kit, a purified methacrylated gelatin kit for UV-crosslinkable hydrogels. PhotoGel® provides 3D tunable gels with the unique attributes to be prepared at various concentrations and crosslinked to provide various gel stiffness.
The PhotoGel® kit consists of purified methacrylated porcine gelatin and a UV Photoinitiator (365 nm). The gelatin is type A 300 bloom.
PhotoGel® is produced from methacrylated gelatin where the gelatin has been modified by reacting the free amines, primarily the ε-amines groups of the lysine residues as well as the a-amines groups on the N-termini. >75% of the total lysine residues of the gelatin molecule have been methacrylated. The photoinitiator consists of Irgacure 2959 to be formulated in methanol (methanol not included) which allows UV crosslinking of the collagen at 365 nm.
Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty