Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PEGSSDA

Supplier: ADVANCED BIOMATRIX, INC. MS
Synonyms: Polyethylene glycol diacrylatePoly(ethylene glycol) diacrylatePEG bisacrylatePEG hydrogelPEGDAPEG diacrylate

GS755-2EA
103792-972PK 209.23 USD InStock
103792-972
PEGSSDA
Polyethylene glycol diacrylate (PEGDA)
A Thiol-reactive crosslinker that allows for easy dissolution of HyStem

  • Use PEGSSDA to allow the non-enzymatic digestion of HyStem hydrogels

PEGSSDA allows researchers to quickly and gently dissolve HyStem® hydrogels non-enzymatically and easily recover cells grown on top or within the hydrogel. PEGSSDA contains two internal disulfide bonds which are rapidly reduced with small amounts of reducing agent causing gel liquefaction.

PEGSSDA allows researchers to quickly and gently dissolve HyStem® hydrogels non-enzymatically, thereby easily recovering cells or spheroids either grown on top or within the hydrogel. PEGSSDA contains two internal disulfide bonds which are rapidly reduced with small amounts of reducing agent (such as N-acetyl cysteine or glutathione), causing gel liquefaction.

PEGSSDA is a pentablock PEGDA molecule composed of four PEG molecules (MW 2000 Da) and two interdigitated disulfide bonds. Only the external PEG molecules are acrylated. The final molecular weight is approximately 8500 Da. Like PEGDA, PEGSSDA is a thiol-reactive crosslinker that covalently reacts with the thiol groups in Glycosil®, HyStem, Heprasil®, and Gelin-S® to form viscoelastic hydrogels. PEGSSDA is lyophilized in water.

Caution: This product is for R&D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.


Formula: CH₂=COC(OCH₂CH₂)nOCOC=CH₂
Storage Temperature: Freezer
MDL Number: MFCD00081876
CAS Number: 26570-48-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty