Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water, sterile filtered, degassed

Supplier: ADVANCED BIOMATRIX, INC. MS
Synonyms: Dihydrogen oxide

GS240-10EA GS241-10EA
103792-944PK 183.08 USD InStock
103792-944 103792-946
Water, sterile filtered, degassed
Water
Sterile filtered, degassed, deionized water at pH 5.0-8.0. This product was designed and intended for use with the HyStem® family of thiolated hyaluronic acid products. 

  • Degassed
  • Sterile Filter
  • Deionized


Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty