Would you like to log in again?
Contec Fetal Doppler
Supplier: L&W OPTICS ELECTRONICS LTD SE
LVA046
470323-834EA 75.49 USD InStock
470323-834
Contec Fetal Doppler
Educational Materials Biology Educational Materials Human Biology Learning Activities
Record fetal heart rates.
  • Delicate and compact design
  • Two 1.5 V batteries (AA not included)
  • Can last more than eight hours for normal use
This pocket fetal doppler is a hand-held fetal heart rate detecting device. The device has a high resolution, color LCD screen which can display the fetal heartbeat waveform and calculate the FHR value.
Ordering information: This item requires AA sized batteries which are not included.
Delivery information: Includes 8 Mhz probe
Caution: For educational and training use only. Not intended for diagnostic usage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty