Would you like to log in again?
VeriSpec® ICP Multi-Element Standard 29, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010860-100N
RCRV010860100NEA 859.76 USD InStock
RCRV010860100N
VeriSpec® ICP Multi-Element Standard 29, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
29 component standard.
10 ppm: Al, Ag, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Ti, U, V, Zn.
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty