Would you like to log in again?
HOBO® Waterproof Shuttle
Supplier: ONSET COMPUTER CORPORATION SE
U-DTW-1
470323-762EA 394.99 USD InStock
470323-762
HOBO® Waterproof Shuttle
Data Loggers
The HOBO® waterproof shuttle provides convenient readout and relaunching of underwater and outdoor HOBO data loggers with an Optic USB interface, and is waterproof to 20 m (66'). This shuttle can also be used as a base station.
Ordering information: Couplers are included for compatibility with the U-series loggers with optical communication, including the UA pendants, U20 water level, U22 water temp Pro, UTBI TidbiT, U23 Pro temp/RH, U24 conductivity and U26 DO data loggers.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty