Would you like to log in again?
Avantor® Hichrom, Female to Female Luer Adapter
Supplier: VWR
Avantor® Hichrom
HI-314
76399-352EA 60.76 USD InStock
76399-352
Avantor® Hichrom, Female to Female Luer Adapter
Fittings and Connectors
This adapter provides us the opportunity to adapt female Luer to female Luer connection to a threaded connection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty