Would you like to log in again?
VeriSpec® ICP Multi-Element Interference Check Solution A, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010657-100N RV010657-500N
RCRV010657100NEA 974.77 USD InStock
RCRV010657100N RCRV010657500N
VeriSpec® ICP Multi-Element Interference Check Solution A, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
14 component standard.
20000 ppm: Cl, Ca
3000 ppm: Ca
2500 ppm: Fe, Na
2000 ppm: C
1000 ppm: Al, Mg, P, K, S
20 ppm: Mo, Ti, M
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty