Would you like to log in again?
VeriSpec® ICP Multi-Element Interference Check Solution B, RICCA Chemical Company
Supplier: RICCA Chemical Company
VeriSpec®
RV010659-100N
RCRV010659100NEA 455.04 USD InStock
RCRV010659100N
VeriSpec® ICP Multi-Element Interference Check Solution B, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
10 component standard.
20 ppm: Cr, Co, Cu, Mn, Ni, V
10 ppm: As, Cd, Se, Cn
5 ppm: Ag
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty