Would you like to log in again?
Citrulline Fluorometric Assay Kit, BioVision
Supplier: BioVision
AB273309-100TEST
76376-662EA 1026.59 USD InStock
76376-662
Citrulline Fluorometric Assay Kit, BioVision
Assays Enzyme Assays
For measuring total Citrulline concentrations.
  • Easy method for the measurement of total Citrulline concentrations in a wide variety of samples
  • Very specific and rapid method
L-Citrulline is a non-proteogenic, semi-essential amino acid. Citrulline is formed either by ornithine carbamoyltransferase or as a by-product of the Nitric oxide synthase (NOS) activity. Citrulline generated in the NOS reaction can be recycled to arginine by the two enzymes argininosuccinate synthetase (ASS) and argininosuccinate lyase (ASL) acting in the Urea Cycle. Citrulline concentrations have been shown to be elevated in patients with mutated ASS or ASL genes. Watermelon is one of the richest sources of Citrulline.
Additionally, Citrulline has been advertised as sports nutrition supplement due to its involvement in Nitric Oxide (NO) synthesis, which helps with vasodilation. , BioVision’s Citrulline Assay Kit provides a rapid, specific, and easy method for the measurement of total Citrulline concentrations in a wide variety of samples. In this enzymatic assay, Citrulline is converted into a series of intermediates, which further reacts with a probe producing a stable fluorescent signal (Ex/Em = 535/587 nm). The kit is simple, easy to perform, sensitive and is high-throughput adaptable. It can detect as low as 2 µm Citrulline in biological samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty