Would you like to log in again?
Hydroxy Acid Oxidase Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB273341-100TEST
76376-670EA 873.77 USD InStock
76376-670
Hydroxy Acid Oxidase Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
For measuring HAO activity in biological samples.
  • The assay can detect as low as 1 µU of HAO activity in biological samples
  • Simple plate-based fluorometric assay for measuring HAO activity in biological samples
Hydroxy Acid Oxidase (HAO) (EC 1.1.3.15), also known as glycolate oxidase is a flavin mononucleotide dependent enzyme that oxidizes alpha hydroxy acid to oxo acid. Glycolate is the primary substrate for HAO, which gets oxidized to glyoxalate. HAO contributes to the general pathway of fatty acid α-oxidation and plays an important role in ethylene glycol poisoning. It is expressed in liver, kidney and pancreas. The most well studied enzyme of this family is the plant glycolate oxidase that has an essential role in photorespiration.
BioVision’s Hydroxy Acid Oxidase Activity Kit is a simple plate-based fluorometric assay for measuring HAO activity in biological samples. HAO oxidizes hydroxy carboxylate, which is accompanied by the release of H₂O₂. H₂O₂ then oxidizes a non-fluorescent probe to a fluorescent product in the presence of enzyme mix via an enzymatic reaction. The assay can detect as low as 1 µU of HAO activity in biological samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty