Would you like to log in again?
cAMP Phosphodiesterase Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB273342-100TEST
76376-674EA 1015.82 USD InStock
76376-674
cAMP Phosphodiesterase Activity Assay Kit (Fluorometric), BioVision
Assays Enzyme Assays
To measure cPDE activity in various sample types.
  • Sensitive and rapid assay
  • Straightforward way to measure cPDE activity in various sample types
Cyclic adenosine-3’,5’-monophosphate (cyclic AMP or cAMP) is a vital second messenger molecule in eukaryotes as well as a relevant signaling effector in bacteria. Intracellular cAMP-mediated signaling cascades control many biological pathways such as glucose and lipid metabolism via activation of protein kinase A and cyclic nucleotide gated ion channels. Intracellular cAMP is synthesized from ATP by the enzyme adenylyl cyclase and is inactivated by hydrolytic cyclic-AMP phosphodiesterase enzymes (cPDEs; EC 3.1.4.53). Inhibition of cPDEs has been shown to impact many biological pathways including cognition, neuronal signaling, inflammation and vascular smooth muscle contractility. Eleven superfamilies of PDEs with varying selectivity for cAMP or other cyclic nucleotides have been identified in mammals.
The predominant phosphodiesterase isoforms involved in the degradation of cAMP are phosphodiesterase 4 (PDE4) family members. There are many unique isoforms of the enzyme, expressed either as products of different genes or through alternative splicing, and their expression levels are cell-type dependent. , BioVision’s cAMP Phosphodiesterase Activity Assay Kit provides a rapid, sensitive and straightforward way to measure cPDE activity in various sample types. In this assay, AMP produced by cPDE activity is metabolized by the enzyme mix, developer mix and converter mix to generate an intermediate compound, which reacts with a probe, yielding a fluorescent signal that can be measured at Ex/Em = 535/587 nm. This assay can detect cPDE activity as low as 0.1 µU per well.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty