Would you like to log in again?
MMP-2 Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB283367-100TEST
76376-682EA 1032.6 USD InStock
76376-682
MMP-2 Inhibitor Screening Kit (Fluorometric), BioVision
Assays Protein Assays
For screening or characterizing MMP-2 inhibitors.
  • Sensitive way for screening
  • studying and characterizing potential inhibitors of MMP-2
  • Quick
Matrix metalloproteinase-2 (MMP-2) is a zinc-dependent endopeptidase family member that is capable of degrading denatured collagen, collagen type IV as well as other extracellular matrix proteins in normal physiological processes such as embryonic development, tissue remodeling and in disease processes including metastasis, atherosclerosis, cardiac dysfunction etc. MMP-2 is widely expressed in almost all cell types. Like most MMP's, MMP-2 is secreted as inactive pro-protein that is activated when cleaved by extracellular proteases. Additionally, MMP-2 can be activated intracellularly via S-Glutathionylation.
BioVision’s MMP-2 Inhibitor Screening Kit provides a quick and sensitive way for screening, studying and characterizing potential inhibitors of MMP-2. In this assay, MMP-2 hydrolyzes a FRET-based MMP-2 substrate and releases the quenched fluorescent group, Mca, which can then be detected fluorometrically at Ex/Em = 325/393 nm. A potent, specific MMP-2 inhibitor is also included in the kit.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty