Would you like to log in again?
Heat Shock Protein Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB284527-100TEST
76376-696EA 874.91 USD InStock
76376-696
Heat Shock Protein Assay Kit (Fluorometric), BioVision
Assays Protein Assays
To measure the activity of an enzyme during heat stress.
  • Simple plate based assay that measures the activity of an enzyme in its native state and after being subjected to heat stress at 45 °C in the presence and absence of an HSP
Thermal stress or elevated temperatures cause proteins to denature thereby resulting in the loss of function leading to disruption in protein homeostasis. All organisms express a class of proteins known as Heat Shock Proteins (HSPs) to circumvent this problem by stabilizing the protein structure during stressful conditions. The name “Heat Shock Protein” comes from the original discovery that the abundance of these proteins is elevated during heat stress. Although HSPs are constitutively expressed and are involved in protein stabilization by ensuring the correct folding of newly synthesized proteins, their abundance is increased during elevated temperatures, high salt concentration, or altered pH. Under such stressful conditions, HPSs bind to proteins thereby providing protection from misfolding and aggregation. HSPs are named according to their molecular weights.
Some of the most widely studied HSPs are HSP60, HSP70 and HSP90 with a molecular mass of 60, 70 and 90 kDa respectively. HSP90 is known to stabilize tumor specific proteins required for tumor growth and thus its inhibitors are being investigated as potential therapeutic targets. , BioVision’s Heat Shock Protein Assay Kit is a simple plate based assay that measures the activity of an enzyme in its native state and after being subjected to heat stress at 45ºC in the presence and absence of an HSP. Protective effect of the HSP is expressed as the % activity of the enzyme heated in the presence of HSP as compared to the activity of the native enzyme.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty