Would you like to log in again?
FAAH1 Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB283381-100TEST
76376-700EA 1083.17 USD InStock
76376-700
FAAH1 Inhibitor Screening Kit (Fluorometric), BioVision
Assays Enzyme Assays
For high-throughput screening of fatty acid Amide Hydrolase inhibitors.
  • Simple, sensitive and rapid
  • Reliable test suitable for high-throughput screening of FAAH1 inhibitorsSensitive, Rapid
Fatty Acid Amide Hydrolase (FAAH; EC: 3.5.1.99), also known as Oleamide Hydrolase or Anandamide Amidohydrolase is a member of the serine hydrolase family of enzymes. It is an integral membrane enzyme that hydrolyzes the endocannabinoid anandamide and related amidated signaling lipids. Endocannabinoids are endogenous lipid ligands that activate the Cannabinoid G-protein coupled Receptors (CB). CB1 and CB2 modulate physiological and behavioral processes such as pain, anti-inflammation etc. In addition to hydrolyzing Endocannabinoids, FAAH regulates other lipid signaling molecules with anti-inflammatory and analgesic properties. Recent study suggests that FAAH may represent an attractive therapeutic target for treatment of pain, inflammation etc.
BioVision’s Human FAAH1 Inhibitor Screening Kit, FAAH1 hydrolyzes a non-fluorescent substrate to generate a fluorescent product, 7-amino-4-methylcoumarin, which can be measured at Ex/Em = 360/465 nm. In the presence of FAAH1 Inhibitor, the reaction is impeded. The assay is high-throughput adaptable and can be completed in less than one hour.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty