Would you like to log in again?
Avantor® Partisphere SIL, HPLC Columns, 5 µm
Supplier: VWR
Avantor® Partisphere
4621-1501 4621-0501
76398-286EA 775.5 USD InStock
76398-286 76398-274
Avantor® Partisphere SIL, HPLC Columns, 5 µm
Chromatography Columns
Avantor® Partisphere® was one of the first commercially available spherical silicas and continues to provide reproducible, high efficiency separations.
  • Spherical 5 µm porous silica
  • Unique void sealing cartridge hardware
  • Convenient and easy to use - hand tightened fittings
  • Increased column lifetime
Stainless steel coulmn hardware.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty