Would you like to log in again?
Thermal Transfer Repositionable Paper Labels, GA International
Supplier: GA International
807656022260 807656022284 807656022192 807656022222 807656022178 807656022147 807656022116 807656022161 807656022277 807656022185 807656022246 807656022239 807656022109 807656022130 807656022154 807656022215 807656022093 807656022253 807656022208 807656022123
76274-730RL 31.69 USD InStock
76274-730 76274-752 76274-740 76274-750 76274-716 76274-738 76274-726 76274-748 76274-736 76274-746 76274-724 76274-734 76274-744 76274-722 76274-754 76274-732 76274-742 76274-720 76274-718 76274-728
Thermal Transfer Repositionable Paper Labels, GA International
Labels
Repositionable Paper Labels are available in a wide assortment of colors to facilitate color-coding and easy identification.
  • Repositionable adhesive
  • Compatible with desktop thermal printers, or industrial thermal printers
  • Available in 1 or 3" core sizes
General-purpose repositionable labels are supplied perforated, and available in white, yellow, orange, red, blue, green apple, pink, lavender, or silver.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty