Would you like to log in again?
Standard Use Piggyback Labels, GA International
Supplier: GA International
807656096223 807656029146 807656096247 807656029153
76273-700RL 114.2 USD InStock
76273-700 76304-450 76273-702 76304-448
Standard Use Piggyback Labels, GA International
Labels
Permanent standard use, thermal-transfer, piggyback paper labels.
  • Perfect for organizing and managing records
  • Multi-layer piggyback design facilitates product tracking and streamlining
  • Printable with all major thermal-transfer printer brands
The labels have a label-on-label, multi-layer design that allows multiple related labels to be printed at once and placed in one location until the smaller labels within are needed to identify subsequent items. Thermal transfer printable, they are ideal for general use in research laboratories, hospitals, and clinical trial facilities, as well as warehouses and shipping/sorting facilities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty