Would you like to log in again?
Laser and Inkjet Glossy Paper Labels, GA International
Supplier: GA International
807656010403 807656010410 807656010397
76272-218PK 58.23 USD InStock
76272-218 76272-216 76272-214
Laser and Inkjet Glossy Paper Labels, GA International
Labels
General use permanent paper labels, for laser and inkjet desktop printers.
  • Easy to customize, with free word templates available
  • Permanent adhesive ensures labels won’t wrinkle or peel off
  • For use with all common brands of laser and inkjet desktop printers
  • Labels won’t jam printers
  • Resistant to gamma irradiation (tested to up to 50 kGy)
Glossy finish labels will adhere to a variety of surfaces, including paper, glass, cardboard, and plastic. Ideal for office use on files, folders, shelves, drawers, boxes, and other containers. Provided in sheet format, the laser and inkjet printable labels will not jam printers, and can be easily customized using free online templates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty