Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avantor® ACE®, Extended Method Development Kits, 5 µm, HPLC/UHPLC Columns

Supplier: VWR
Avantor® ACE®
MDKE-5-1502U MDKE-5-1003U MDKE-5-2503U MDKE-5-2502U MDKE-5-0503U MDKE-5-1503U MDKE-5-1046U MDKE-5-0502U MDKE-5-1546U MDKE-5-2546U MDKE-5-0546U MDKE-5-1002U
76397-062EA 1166.01 USD InStock
76397-062 76397-086 76397-088 76397-044 76397-046 76397-100 76397-048 76397-102 76397-058 76397-104 76397-090 76397-060
Avantor® ACE®, Extended Method Development Kits, 5 µm, HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.

  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1.7 and 5 µm ) and complementary phases
  • Analytical IDs in a wide range of column lengths
  • Ideal for method development
  • Compatible with all manufacturers' LC systems

A complete range of kits for all requirements is available, including porous, solid core, HILIC and bioanalytical 300 Å column kits. These kits group together the essential column chemistries for method development and are available in a wide range of column formats.

Maximum recommended pH: 2.0 to 8.0 (1.5 to 11.5 for SuperC18)

Max. pressure: 1000 bar

Delivery information: Avantor® ACE® Extended Method Development Kits contain Avantor® ACE® SuperC18™, Avantor® ACE® C18-Amide and Avantor® ACE® CN-ES stainless steel columns.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty