Would you like to log in again?
Vernier Go Direct® Centripetal Force Apparatus
Supplier: Vernier
GDX-CFA
470315-118EA 324.99 USD InStock
470315-118
Vernier Go Direct® Centripetal Force Apparatus
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
The Go Direct Centripetal force apparatus and Go Direct force and acceleration sensor make an ideal combination to explore rotational dynamics.
  • Investigate circular motion by measuring centripetal acceleration in relation to the radius of rotation and angular velocity
  • Explore rotational dynamics through an investigation of the force required to keep a mass in circular motion
  • Explore Newton's second law, angular acceleration, and the moment of inertia as they relate to circular motion
When students use this apparatus with a Go Direct force and acceleration sensor, they can conduct a variety of rotational dynamics investigations with a single experiment setup. This combination easily measures angular velocity and centripetal force and acceleration without the hassle of wires. Go Direct sensors connect directly to mobile device, Chromebook™, or computer using our free Graphical Analysis™ 4 app - no additional equipment or software purchases are necessary.
Ordering information: Go Direct force and acceleration sensor not included. It includes base with adjustable feet, bearing and spindle, rotating beam, sensor bracket, mass carriage with thumb screws, mass carriage with bearings, 50 g masses (2), 100 g masses (4).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty