Would you like to log in again?
Vernier® Flash Photolysis Spectrometer
Supplier: VERNIER SOFTWARE & TECH SE
VSP-FP
470320-040EA 5998.99 USD InStock
470320-040
Vernier® Flash Photolysis Spectrometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
The Vernier Flash photolysis spectrometer is a simple, user-friendly device for demonstrating the fundamental principles of chemical kinetics and photochemistry to undergraduate chemistry students.
  • Spectral coverage of 450 to 750 nm
  • 100 microseconds temporal resolution
  • Spectral resolution determined by the interference filter used (10 nm with the included 600 nm interference filter)
  • 15 milliseconds minimum time window
Photochemical reactions are very important in many areas of chemistry with applications including photodynamic therapy and solar energy harvesting. These reactions often proceed quickly and, as a result, require fast reaction techniques to analyze. This is a simple, user-friendly transient absorption spectrometer. It is useful for demonstrating the fundamental principles of these types of fast, photo-catalyzed reactions. Some experiment ideas include kinetics of photochemical reactions such as: Excited-state dynamics, Triplet-decay analysis, Time-resolved phosphorescence, Phosphorescence-quenching kinetics, Isomerization, Reactive free radicals, and Photocatalysis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty