Would you like to log in again?
Scott/Cross-Flow Sample Introduction Torch Module for Optima™ 2x00/4x00/5x00/7x00 DV, PerkinElmer
Supplier: PerkinElmer
N0770606
10136-310EA 4036.83 USD InStock
10136-310
Scott/Cross-Flow Sample Introduction Torch Module for Optima™ 2x00/4x00/5x00/7x00 DV, PerkinElmer
Atomizers
This quick-change sample introduction module, with Scott spray chamber and GemTip Crossflow nebulizer technologies, includes core components and consumables used with Optima™ 2x00 DV, 4x00 DV, 5x00 DV, and 7x00 DV ICP-OES series instruments.
This module kit includes: (1) Inner Chamber for External Ryton/Type II Spray Chamber, (1) Outer Axial Spray Chamber, (1) One-Slot Quartz Torch, (1) GemTip Cross-Flow II Nebulizer, (1) 2.0 mm I.D. Alumina Injector, (1) Quick-Change Torch Module Mounting Block, and (1) Scott Spray Chamber External Drain Kit.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty