Would you like to log in again?
Mosquito Feeders, Large, 50 mm Ø Feeding Area, Membrane Style, GlassChemglass
Supplier: Chemglass
CG-1836-75
76333-626EA 297.63 USD InStock
76333-626
Mosquito Feeders, Large, 50 mm Ø Feeding Area, Membrane Style, GlassChemglass
Animal Supplies Animal Environmental Enrichment
Glass mosquito feeder, Similar to CG-1835, but with a larger feeding surface.
Mosquito feeding area is 50 mm diameter. The outer chamber is for circulating 37 °C water and an inner chamber into which the blood is injected. Parafilm™ is stretched over the lower portion of the inner chamber. The assembled feeder is held on top of the screened box. Feeder has 10 mm OØ hose connections, while the blood chamber has an OØ of 56 mm where the Parafilm™ is installed.
Ordering information: PLEASE NOTE – Feeder is glass only, NOT supplied with Parafilm™ or container.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty