Would you like to log in again?
Adapter, Process Reactor, Lower Outlet, Chemglass
Supplier: Chemglass
CG-1964-50A
76333-374EA 89.22 USD InStock
76333-374
Adapter, Process Reactor, Lower Outlet, Chemglass
Fittings and Connectors
Glass filled PTFE adapter for use with the CG-1968-P zero dead space detachable valve.
Adapts 1" beaded pipe to a ³/₄" compression fitting for use with CG-1975 adapters and tubing.
Ordering information: Requires a CG-1968-66 1" beaded pipe coupling.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty