Would you like to log in again?
Support/Holding Rods, PT Sensors, Hot Plate, Chemglass
Supplier: Chemglass
CG-1995-V-27
76333-662EA 43.84 USD InStock
76333-662
Support/Holding Rods, PT Sensors, Hot Plate, Chemglass
Supports Flask Supports
For fastening a temperature sensor/probe with a boss head to the vertical support rod on a hotplate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty