Would you like to log in again?
Adapters, Air Inlets/Outlets, for Bioreactors, Chemglass
Supplier: Chemglass
CLS-1380-55 CLS-1380-18 CLS-1385-27
76333-446EA 224.65 USD InStock
76333-446 76333-442 76333-460
Adapters, Air Inlets/Outlets, for Bioreactors, Chemglass
Fittings and Connectors
Adapters, air inlets/outlets, overlay aeration, for bioreactors.
Adapter can be used as a gas outlet pipe in lieu of a condenser in applications where the aeration rate or temperature is low or for short term batch applications. more often, this pipe is used to provide overlay aeration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty