Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride 0.9% USP

Supplier: NURSE ASSIST, INC.
Synonyms: HaliteSea saltSaline

6281 6240 6280 6270 6220
76285-236CS 65.74 USD InStock
76285-236 76285-248 76285-242 76285-234 76285-244
Sodium chloride 0.9% USP
Sodium chloride
For irrigation and suction cather prodocedures.

  • for wounds


Formula: NaCl
MW: 58.44 g/mol
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty