Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Transverse Cutting Tweezers, Excelta®

Supplier: Excelta

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

15
75880-530EA 188.98 USD InStock
75880-530
Transverse Cutting Tweezers, Excelta®
Forceps
These cutting tweezers are recommended for cutting 0.010" (0.25 mm) or smaller wire.

Tweezers with epoxy coating have a surface resistivity of 10⁵ to 10⁶ ohms/sq.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty