Would you like to log in again?
Eye Loups, Excelta®
Supplier: Excelta
405 422 410
75880-602EA 66.67 USD InStock
75880-602 75880-604 75880-606
Eye Loups, Excelta®
Magnifiers
Eye loups feature a two element, optical glass construction.
  • Bright, sharp focus loups
  • Wide range of views
Eye loups have 360° clear acrylic bases to admit light from all directions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty