Would you like to log in again?
Leica CM1520 Cryostat for Routine Histology and Mohs Surgery
Supplier: Leica
149491520US 149491520U1
76404-228EA 38881.01 USD InStock
76404-228 76404-230
Leica CM1520 Cryostat for Routine Histology and Mohs Surgery
Microtomes and Microtome Assemblies Microtomes
The Leica CM1520 is a value-priced cryostat for cryosectioning including critical applications such as Mohs surgery.
  • Delivers high-quality sections in a fast, simple and efficient way
Time is crucial when sectioning frozen specimens. An actively cooled quick freezing shelf with defrost function and a powerful refrigeration system are provided to get rapid results.
Less maintenance - more consistent sections: It's easy to consistently create high-quality sections with the Leica CM1520's precise microtome and stepper motor control. The microtome is completely encapsulated, saving valuable time with maintenance-free reliability.
Well organized work space: Create the ideal work environment you've always wanted. The flexible and efficient Leica CM1520 workspace includes a new object plate holder, a movable shelf and designated slide slots and tool storage areas on top of the instrument.
Easy to learn: Self-explanatory, intuitive, direct controls allow anyone to start working right away without long hours of training. No need to scroll through multiple long menu options. Save time by directly setting the function you need.
Fast orientation: The object head orientation, with zero-position centering, enables the specimen to be approached from the desired angle. Its easy handling helps you accomplish your work before deadlines are due.
Ordering information: For available options, configurations, and accessories (sold separately), contact your VWR representative.
Delivery information: Leica Premium CM1520 Mohs model features freestanding cryostat with encapsulated, splash-proof microtome; spacious, stainless-steel cryochamber with antiglare illumination; easy to clean and disinfect; heated, removable sliding window; stable, self-contained cryocabinet on casters; handwheel manually lockable in two positions, and premium CE blade holder.
Leica CM1520 configuration 1 model features freestanding cryostat with encapsulated, splash-proof microtome; spacious, stainless-steel cryochamber with antiglare illumination; easy to clean and disinfect; heated, removable sliding window; stable, self-contained cryocabinet on casters; handwheel manually lockable in two positions and standard CE blade holder.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty