Would you like to log in again?
MicroMist™ Nebulizer for ELAN/NexION, PerkinElmer
Supplier: PerkinElmer
N0775342
10140-322EA 2506.81 USD InStock
10140-322
MicroMist™ Nebulizer for ELAN/NexION, PerkinElmer
Atomizers
This glass MicroMist™ nebulizer includes connectors and sample tubing for use with ELAN® 5000/6x00/9000/DRCs and NexION® 300 and 350 ICP-MS series instruments. It can also be used with Optima® 3x00 RL ICP-OES series instruments.
The MicroMist™ micro-uptake glass concentric nebulizer offers high performance for limited sample volumes. MicroMist™ for ELAN/NexION 300/350 are fitted with a UniFit connector with 1.3×0.50×700 (I.Ø×O.Ø×L) mm sample tubing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty